Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

244597

Sigma-Aldrich

Gold(III) chloride trihydrate

ACS reagent, ≥49.0%

Synonym(s):

Hydrogen tetrachloroaurate(III), Tetrachloroauric(III) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4 · 3H2O
CAS Number:
Molecular Weight:
393.83
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

concentration

≥49.0%

impurities

≤0.1% insol. Ether

cation traces

alkalies and other metals: ≤0.2%

SMILES string

Cl[H].[H]O[H].[H]O[H].[H]O[H].Cl[Au](Cl)Cl

InChI

1S/Au.4ClH.3H2O/h;4*1H;3*1H2/q+3;;;;;;;/p-3

InChI key

XYYVDQWGDNRQDA-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used in preparation of stable nanosized and underivatized colloidal gold particles in solution.[1]

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

target_organs

Kidney

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service