About This Item
Skip To
SMILES string
[Li+].CC(C)(C)O[AlH-](OC(C)(C)C)OC(C)(C)C
InChI
1S/3C4H9O.Al.Li.H/c3*1-4(2,3)5;;;/h3*1-3H3;;;/q3*-1;+2;+1;
InChI key
BYBIDFZFKATBFH-UHFFFAOYSA-N
reaction suitability
reagent type: reductant
concentration
0.5 M in 2-methoxyethyl ether
density
0.942 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
Application
- Reducing agent
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1B - Water-react 1
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk
WGK 1
flash_point_f
95.0 °F - closed cup
flash_point_c
35 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


