245992

Sigma-Aldrich

4-Aminophenol

technical grade, 95%

Synonym(s):
4-Hydroxyaniline
Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein/REAXYS Number:
385836
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

assay

95%

mp

185-189 °C (lit.)

SMILES string

Nc1ccc(O)cc1

InChI

1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

PLIKAWJENQZMHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Y Xu et al.
Clinical chemistry, 36(11), 1941-1944 (1990-11-01)
A sandwich-type heterogeneous enzyme immunoassay with flow-injection analysis for alpha-fetoprotein (AFP) in human serum has been developed. 4-Aminophenol, the product of enzymatic reaction, is detected amperometrically. The interassay CV for this electrochemical enzyme immunoassay was less than 8.2%, with a...
E P Gil et al.
Clinical chemistry, 36(4), 662-665 (1990-04-01)
A competitive enzyme-linked immunoabsorbent assay based on the flow-injection amperometric detection of p-aminophenol has been investigated with use of the materials and general procedure of a commercial kit for the determination of theophylline in human serum. The antibody is immobilized...
M Meusel et al.
Biosensors & bioelectronics, 10(6-7), 577-586 (1995-01-01)
Apolipoprotein (apo) E is a constituent of serum lipoproteins and can serve as a diagnostic parameter for the assessment of disorders of lipid metabolism. For the determination of apo E in serum a sandwich-type amperometric immunosensor using disposable membranes was...
Tomoyuki Yasukawa et al.
Biosensors & bioelectronics, 37(1), 19-23 (2012-05-23)
In this work, a novel immunosensing method has been developed on the basis of the sensitive determination of a product generated by an enzyme reaction with dual amplification system combining an electrochemical-redox cycling and coulometric signal transduction using a galvanic...
Solomon Mehretie et al.
Talanta, 85(3), 1376-1382 (2011-08-03)
A sensitive and selective method was developed for the determination of N-acetyl-p-aminophenol (APAP) and p-aminophenol (PAP) using poly(3,4-ethylenedioxythiophene) (PEDOT)-modified glassy carbon electrode (GCE). Cyclic voltammetry and differential pulse voltammetry were used to investigate the electrochemical reaction of APAP and PAP...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service