Documents

249122

trans-11-Tetradecenyl acetate

97%

Synonym(s):
(E)-11-TDA, E-11-TDA
Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
MDL number:

assay

97%

SMILES string

CC\C=C\CCCCCCCCCCOC(C)=O

InChI

1S/C16H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h4-5H,3,6-15H2,1-2H3/b5-4+

InChI key

YJINQJFQLQIYHX-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service