250902

Sigma-Aldrich

1-Ethylnaphthalene

≥97%

Linear Formula:
C10H7C2H5
CAS Number:
Molecular Weight:
156.22
Beilstein/REAXYS Number:
1854417
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

≥97%

refractive index

n20/D 1.606 (lit.)

bp

258-260 °C (lit.)

mp

−15-−14 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

SMILES string

CCc1cccc2ccccc12

InChI

1S/C12H12/c1-2-10-7-5-8-11-6-3-4-9-12(10)11/h3-9H,2H2,1H3

InChI key

ZMXIYERNXPIYFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Ethylnaphthalene has been employed as guest molecule to investigate:
  • effect of binding Tb3+ to sodium taurocholate aggregates containing polyaromatic hydrocarbon guests
  • its binding to aggregates of sodium cholate, taurocholate, deoxycholate and deoxytaurocholate

Packaging

5 g in glass bottle

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup

Rui Li et al.
Langmuir : the ACS journal of surfaces and colloids, 25(24), 13800-13808 (2009-07-18)
The binding of naphthalene (Np), 1-ethylnaphthalene (EtNp), acenaphthene (AcN), and 1-naphthyl-1-ethanol (NpOH) as guests to the aggregates of sodium cholate (NaCh), taurocholate (NaTC), deoxycholate (NaDC), and deoxytaurocholate (NaTDC) was studied with the objective of determining how the structure of the...
Tamara C S Pace et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(10), 1568-1577 (2011-07-14)
The effect of binding Tb(3+) to sodium taurocholate aggregates containing polyaromatic hydrocarbon guests was examined using pyrene and 1-ethylnaphthalene as guests that bind to the primary aggregate, and 1-naphthyl-1-ethanol as a secondary aggregate guest. Time-resolved fluorescence quenching studies were used...
Hyun-Joong Kang et al.
Chemosphere, 222, 656-664 (2019-02-08)
Crude oil released into the environment contains many polycyclic aromatic hydrocarbons (PAHs). Alkylated PAHs are more abundant than unsubstituted PAHs and their toxicity is also of serious concern. Among the various physical, chemical, and biological weathering processes of crude oils...
Danlei Wang et al.
Chemico-biological interactions, 315, 108905-108905 (2019-11-26)
Mineral oils are widely applied in food production and processing and may contain polycyclic aromatic hydrocarbons (PAHs). The PAHs that may be present in mineral oils are typically alkylated, and have been barely studied. Metabolic oxidation of the aromatic ring...
Nien-Hsin Kao et al.
Marine pollution bulletin, 97(1-2), 319-332 (2015-06-08)
Three fishing harbors were investigated to study the polycyclic aromatic hydrocarbons in the sediments and trace possible anthropogenic sources by identification of cyclic terpenoid biomarkers. Seventeen terpanes, 10 steranes and 10 bicyclic sesquiterpanes in the marine diesel and the three...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.