Documents

251461

Sigma-Aldrich

N-(3,5-Dinitrobenzoyl)-DL-α-phenylglycine

98%

Linear Formula:
(O2N)2C6H3CONHCH(C6H5)CO2H
CAS Number:
Molecular Weight:
345.26
MDL number:

assay

98%

mp

240 °C (dec.) (lit.)

SMILES string

OC(=O)C(NC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O)c2ccccc2

InChI

1S/C15H11N3O7/c19-14(16-13(15(20)21)9-4-2-1-3-5-9)10-6-11(17(22)23)8-12(7-10)18(24)25/h1-8,13H,(H,16,19)(H,20,21)

InChI key

MIVUDAUOXJDARR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service