252085

Sigma-Aldrich

9,10-Dichloroanthracene

98%

Empirical Formula (Hill Notation):
C14H8Cl2
CAS Number:
Molecular Weight:
247.12
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

213-215 °C (lit.)

SMILES string

Clc1c2ccccc2c(Cl)c3ccccc13

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service