MilliporeSigma
All Photos(1)

253138

Sigma-Aldrich

9-BBN triflate solution

0.5 M in hexanes

Synonym(s):
9-Borabicyclo[3.3.1]nonyl trifluoromethanesulfonate solution, 9-BBN triflate solution, 9-BBN·CF3SO3, 9-[[(Trifluoromethyl)sulfonyl]oxy]-9-borabicyclo[3.3.1]nonane
Empirical Formula (Hill Notation):
C9H14BF3O3S
CAS Number:
Molecular Weight:
270.08
Beilstein:
995656
MDL number:
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

0.5 M in hexanes

density

0.733 g/mL at 25 °C

SMILES string

FC(F)(F)S(=O)(=O)OB1C2CCCC1CCC2

InChI

1S/C9H14BF3O3S/c11-9(12,13)17(14,15)16-10-7-3-1-4-8(10)6-2-5-7/h7-8H,1-6H2/t7-,8+

InChI key

NXHUKDCGEGEZCX-OCAPTIKFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant for:
  • Preparation of a dialkylborenium ion stabilized by an N-heterocyclic carbene
  • Mukaiyama aldol addition
  • Preparation of porphyrin dimethyldioxanyl tolyl derivative complex
  • Preparation of palladium porphyrin complexes
  • Boron-mediated β-alkoxy Me ketone aldol addition reactions

  • Enolization agent

Packaging

100 mL in Sure/Seal™

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service