MilliporeSigma
All Photos(1)

256048

Sigma-Aldrich

Vanadium(V) oxychloride

99.995% trace metals basis

Synonym(s):
Vanadyl trichloride, Vanadium(V) oxytrichloride, Trichlorooxovanadium oxide
Linear Formula:
VOCl3
CAS Number:
Molecular Weight:
173.30
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

13.8 mmHg ( 20 °C)

assay

99.995% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: vanadium

bp

126-127 °C (lit.)

mp

−77 °C (lit.)

density

1.84 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[V](Cl)(Cl)=O

InChI

1S/3ClH.O.V/h3*1H;;/q;;;;+3/p-3

InChI key

JBIQAPKSNFTACH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Masayuki Kirihara et al.
Chemical & pharmaceutical bulletin, 52(5), 625-627 (2004-05-11)
Thiols were converted into disulfide by the aerobic oxidation catalyzed by trichlorooxyvanadium in the presence of molecular sieves 3A.
[Hygienic characteristics of the working conditions in the manufacture of vanadium hydroxychloride].
B G Feĭgin et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 51-52 (1985-01-01)
M D Zidan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(14), 2693-2698 (2001-01-06)
A new route has been devised, leading to the production of VOX3 molecules where X=F, Br and I by an on-line process using vanadium oxytrichloride, VOCl3 as a starting compound passed over the following heated salts NaF, KBr and KI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service