257141

Sigma-Aldrich

Sodium diethyldihydroaluminate solution

25 wt. % in toluene

Synonym(s):
OMH-1, Sodium aluminum diethyl dihydride
Empirical Formula (Hill Notation):
C4H12AlNa
CAS Number:
Molecular Weight:
110.11
MDL number:

concentration

25 wt. % in toluene

impurities

5% tetrahydrofuran

SMILES string

[Na+].CC[AlH2-]CC

InChI

1S/2C2H5.Al.Na.2H/c2*1-2;;;;/h2*1H2,2H3;;;;/q;;-1;+1;;

InChI key

DJNBMRGHGJSTBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under nitrogen in Sure/Pac™ metal cylinder

Signal Word

Danger

hazcat

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

storage_class_code

3 - Flammable liquids

WGK Germany

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service