Documents

257516

Sigma-Aldrich

Ethyl triacontanoate

98%

Synonym(s):
Melissic acid ethyl ester
Linear Formula:
CH3(CH2)28CO2C2H5
CAS Number:
Molecular Weight:
480.85
MDL number:

assay

98%

mp

67-69 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OCC

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service