258733

Sigma-Aldrich

Heptanoic acid

99%

Synonym(s):
Enanthic acid, Oenanthic acid
Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
Beilstein/REAXYS Number:
1744723
EC Number:
MDL number:

vapor density

4.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

assay

99%

expl. lim.

10.1 %

refractive index

n20/D 1.4221 (lit.)

bp

223 °C (lit.)

mp

−10.5 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(O)=O

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Christian Pellevoisin et al.
Toxicology in vitro : an international journal published in association with BIBRA, 50, 418-425 (2018-01-18)
According to ISO 10993 standards for biocompatibility of medical devices, skin irritation is one of the three toxicological endpoints to be always addressed in a biological risk assessment. This work presents a new protocol to assess this endpoint in vitro...
Helena Kandarova et al.
Toxicology in vitro : an international journal published in association with BIBRA, 50, 407-417 (2018-02-14)
Assessment of dermal irritation is an essential component of the safety evaluation of medical devices. Reconstructed human epidermis (RhE) models have replaced rabbit skin irritation testing for neat chemicals and their mixtures (OECD Test Guideline 439). However, this guideline cannot...
Gregory Kaler et al.
The Journal of biological chemistry, 282(33), 23841-23853 (2007-06-08)
Organic anion transporters (OATs, SLC22) interact with a remarkably diverse array of endogenous and exogenous organic anions. However, little is known about the structural features that determine their substrate selectivity. We examined the substrate binding preferences and transport function of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service