All Photos(1)

258873

Sigma-Aldrich

Ethyl 2-methylcyclopropanecarboxylate

96%

Empirical Formula (Hill Notation):
C7H12O2
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
PubChem Substance ID:

assay

96%

SMILES string

CCOC(=O)C1CC1C

InChI

1S/C7H12O2/c1-3-9-7(8)6-4-5(6)2/h5-6H,3-4H2,1-2H3

InChI key

JWYSLVLBKXDZCW-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service