259985

Sigma-Aldrich

Methyl 2-methylbenzoate

99%

Synonym(s):
Methyl o-toluate
Linear Formula:
CH3C6H4CO2CH3
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

99%

refractive index

n20/D 1.519 (lit.)

bp

207-208 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1C

InChI

1S/C9H10O2/c1-7-5-3-4-6-8(7)9(10)11-2/h3-6H,1-2H3

InChI key

WVWZECQNFWFVFW-UHFFFAOYSA-N

Related Categories

General description

Efficiency of reduction of methyl 2-methylbenzoate using different heterogeneous systems has been compared.

Application

Methyl 2-methylbenzoate has been used in the preparation of 2-(azidomethyl)benzoic acid.

Packaging

25 g in glass bottle

Certificate of Analysis

Certificate of Origin

2-(Azidomethyl) benzoyl as a new protecting group in nucleosides.
Wada T, et al.
Tetrahedron Letters, 42(6), 1069-1072 (2001)
A catalytic stereo-and chemo-selective method for the reduction of substituted aromatics.
Fache F, et al.
Tetrahedron Letters, 36(6), 885-888 (1995)
Nicholas R Larson et al.
Journal of medical entomology, 57(1), 187-191 (2019-09-10)
Common bed bug Cimex lectularius (L.) (Hemiptera: Cimicidae) infestations are on the rise and due to the development of pesticide resistance they are becoming more difficult to control, affordably. We evaluated a naturally occurring compound methyl benzoate (MB) and related...
Yan Feng et al.
Scientific reports, 8(1), 7902-7902 (2018-05-23)
Benzyl methyl ester, also known as methyl benzoate (MB), is a volatile organic compound that exists naturally as a floral fragrance in many plants. Our behavioral bioassays show that MB and some of its naturally occurring and synthetic analogs kill...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service