Documents

260800

Sigma-Aldrich

Indophenol sodium salt

suitable as redox indicator

Synonym(s):
4-[(4-Hydroxyphenyl)imino]-2,5-cyclohexadien-1-one sodium salt
Empirical Formula (Hill Notation):
C12H8NNaO2
CAS Number:
Molecular Weight:
221.19
Beilstein/REAXYS Number:
4611557
MDL number:
PubChem Substance ID:

form

powder

application(s)

hematology: suitable
histology: suitable

solubility

1 M NaOH: 10 mg/mL

Featured Industry

Diagnostic Assay Manufacturing

storage temp.

room temp

SMILES string

[Na+].[O-]c1ccc(cc1)\N=C2\C=CC(=O)C=C2

InChI

1S/C12H9NO2.Na/c14-11-5-1-9(2-6-11)13-10-3-7-12(15)8-4-10;/h1-8,14H;/q;+1/p-1

InChI key

FRUIYGAEJNTDCA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Certificate of Origin

Hyun-A Nam et al.
Food chemistry, 286, 338-345 (2019-03-05)
Consumption and global utilization of mandarins is on the rise due to its easy-to-peel attribute compared to other citrus fruits. Mandarins are irradiated as an effective phyto-sanitary treatment for trade requirements. In this study, mandarins were irradiated with 0.4 and...
Ricardo Pedregal-Cortés et al.
Biofouling, 35(7), 732-741 (2019-08-31)
Silk sericin is a globular protein whose resistance against fouling is important for applications in biomaterials and water-purification membranes. Here it is shown how sericin generates a water-exclusion zone that may facilitate antifouling behavior. Negatively charged microspheres were used to...
Sheemona Chowdhary et al.
Biochimie, 154, 194-209 (2018-09-12)
Erythrocyte acetylcholinesterase (AChE) is a preferred biomarker for the detection of organophosphorus poisoning. Acetylthiocholine (ATCh) is the most popular substrate for the detection of AChE activity. However, oximolysis is a prominent feature with ATCh. In this context, we have searched...
Cole D Libner et al.
The Journal of comparative neurology, 528(10), 1704-1724 (2019-12-25)
Neurodegeneration, including loss of neurons and axons, is a feature of progressive forms of multiple sclerosis (MS). The mechanisms underlying neurodegeneration are mostly unknown. Research implicates autoimmunity to nonmyelin self-antigens as important contributors to disease pathogenesis. Data from our lab...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service