264822

Sigma-Aldrich

Phenyl tributyltin sulfide

95%

Linear Formula:
[CH3(CH2)3]3SnSC6H5
CAS Number:
Molecular Weight:
399.22
MDL number:

assay

95%

refractive index

n20/D 1.544 (lit.)

bp

147 °C/0.3 mmHg (lit.)

density

1.172 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)Sc1ccccc1

Certificate of Analysis

Certificate of Origin

Aldrichimica Acta, 17, 26-26 (1984)
Tetrahedron Letters, 2853-2853 (1979)
Aldrichimica Acta, 17, 75-75 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service