266663
99.998% trace metals basis
rod
6.24 μΩ-cm, 20°C
5.0 mm
2900 °C (lit.)
8.9 g/mL at 25 °C (lit.)
[Co]
1S/Co
GUTLYIVDDKVIGB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | GF01747403 | GF00415426 | GF00377029 |
---|---|---|---|
assay 99.998% trace metals basis | assay 99.90% | assay ≥99.9% | assay ≥99.9% |
form rod | form foil | form foil | form foil |
resistivity 6.24 μΩ-cm, 20°C | resistivity 6.24 μΩ-cm, 20°C | resistivity 6.24 μΩ-cm, 20°C | resistivity 6.24 μΩ-cm, 20°C |
bp 2900 °C (lit.) | bp 2900 °C (lit.) | bp 2900 °C (lit.) | bp 2900 °C (lit.) |
density 8.9 g/mL at 25 °C (lit.) | density 8.9 g/mL at 25 °C (lit.) | density 8.9 g/mL at 25 °C (lit.) | density 8.9 g/mL at 25 °C (lit.) |
diam. 5.0 mm | diam. - | diam. - | diam. - |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service