All Photos(1)

267694

Sigma-Aldrich

Zirconium

wire, diam. 0.127 mm, 99.95% trace metals basis

Synonym(s):
Zirconium element
Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
EC Number:
MDL number:
PubChem Substance ID:

assay

99.95% trace metals basis

form

wire

resistivity

40 μΩ-cm, 20°C

diam.

0.127 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Thijs H Oude Munnink et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 929-935 (2013-04-30)
Placental growth factor (PlGF) is a member of the proangiogenic vascular endothelial growth factor family, which is upregulated in many tumors. RO5323441, a humanized monoclonal antibody against PlGF, showed antitumor activity in human tumor xenografts. We therefore aimed to radiolabel...
O Tymofiyeva et al.
Dento maxillo facial radiology, 42(6), 20120271-20120271 (2013-04-24)
To investigate the potential influence of standard dental materials on dental MRI (dMRI) by estimating the magnetic susceptibility with the help of the MRI-based geometric distortion method and to classify the materials from the standpoint of dMRI. A series of...
Surgical, biologic and implant-related factors affecting bone remodeling around implants.
Matteo Capelli
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry, 8(2), 279-313 (2013-05-29)
New CAD/CAM materials and blocks for chairside procedures.
M Zimmermann et al.
International journal of computerized dentistry, 16(2), 173-181 (2013-08-13)
[New bipolar electrosurgical tools based on zirconium dioxide].
S V Belov et al.
Meditsinskaia tekhnika, (2)(2), 20-24 (2013-06-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service