267910

Sigma-Aldrich

Zinc

wire, diam. 0.25 mm, 99.99%

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:

vapor pressure

1 mmHg ( 487 °C)

assay

99.99%

form

wire

resistivity

5.8 μΩ-cm, 20°C

diam.

0.25 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service