All Photos(1)

268631

Sigma-Aldrich

Bis(salicylaldehyde)cobalt(II) dihydrate

97%

Linear Formula:
[HCOC6H4-2-(O-)]2Co·2H2O
CAS Number:
Molecular Weight:
337.19
MDL number:
PubChem Substance ID:

assay

97%

reaction suitability

reagent type: catalyst
core: cobalt

SMILES string

O.O.O=Cc1ccccc1O[Co]Oc2ccccc2C=O

InChI

1S/2C7H6O2.Co.2H2O/c2*8-5-6-3-1-2-4-7(6)9;;;/h2*1-5,9H;;2*1H2/q;;+2;;/p-2

InChI key

UJABLPZHFYKNIW-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service