Documents

272043

Sigma-Aldrich

3,3′-Bis(trifluoromethyl)benzophenone

98%

Linear Formula:
(CF3C6H4)2CO
CAS Number:
Molecular Weight:
318.21
MDL number:
PubChem Substance ID:

assay

98%

mp

100-102 °C (lit.)

SMILES string

FC(F)(F)c1cccc(c1)C(=O)c2cccc(c2)C(F)(F)F

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service