Skip to Content
MilliporeSigma

274151

3-Methyl-1-penten-3-ol

99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
100.16
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
213-044-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

refractive index

n20/D 1.428 (lit.)

bp

117-118 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(O)C=C

InChI

1S/C6H12O/c1-4-6(3,7)5-2/h4,7H,1,5H2,2-3H3

InChI key

HFYAEUXHCMTPOL-UHFFFAOYSA-N

General description

The reactions of 3-methyl-1-penten-3-ol with ozone and OH radicals was studied using simulation chambers and the rate co-efficient values were determined. The hydrogenation of 3-methyl-1-penten-3-ol was also studied.


Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

WGK

WGK 3

Flash Point (°F)

78.8 °F - closed cup

Flash Point (°C)

26 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library