MilliporeSigma
All Photos(1)

276081

Sigma-Aldrich

(R,R)-(+)-N,N′-Bis(α-methylbenzyl)sulfamide

99%

Linear Formula:
[C6H5CH(CH3)NH]2SO2
CAS Number:
Molecular Weight:
304.41
MDL number:
PubChem Substance ID:

Quality Level

assay

99%

optical activity

[α]20/D +80°, c = 2 in ethanol

mp

98-100 °C (lit.)

SMILES string

C[C@@H](NS(=O)(=O)N[C@H](C)c1ccccc1)c2ccccc2

InChI

1S/C16H20N2O2S/c1-13(15-9-5-3-6-10-15)17-21(19,20)18-14(2)16-11-7-4-8-12-16/h3-14,17-18H,1-2H3/t13-,14-/m1/s1

InChI key

ZUMVQOILJDLACR-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service