277274

Sigma-Aldrich

4-Nitrophenyl 3-bromo-2,2-diethoxypropionate

97%

Linear Formula:
BrCH2C(OC2H5)2CO2C6H4NO2
CAS Number:
Molecular Weight:
362.17
MDL number:

assay

97%

mp

75-78 °C (lit.)

SMILES string

CCOC(CBr)(OCC)C(=O)Oc1ccc(cc1)[N+]([O-])=O

Application

Starting material for heterocycles.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service