Documents

280461

2,6-Di-tert-butylnaphthalene

technical grade

Empirical Formula (Hill Notation):
C18H24
CAS Number:
Molecular Weight:
240.38
EC Number:
MDL number:

grade

technical grade

SMILES string

CC(C)(C)c1ccc2cc(ccc2c1)C(C)(C)C

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service