Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H12NNaO8S2 · xH2O
CAS Number:
Molecular Weight:
445.40 (anhydrous basis)
PubChem Substance ID:
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
4100175
MDL number:
assay
90%
form
powder
solubility
NaOH: soluble 5%, clear to hazy, yellow to brown
SMILES string
O.[Na+].Oc1ccccc1\C=N\c2cc(cc3cc(cc(O)c23)S(O)(=O)=O)S([O-])(=O)=O
InChI
1S/C17H13NO8S2.Na.H2O/c19-15-4-2-1-3-10(15)9-18-14-7-12(27(21,22)23)5-11-6-13(28(24,25)26)8-16(20)17(11)14;;/h1-9,19-20H,(H,21,22,23)(H,24,25,26);;1H2/q;+1;/p-1/b18-9+;;
InChI key
AAIGDXDVSZJVSW-IFJQNBRBSA-M
General description
The tautomeric equillibrium of azomethine-H monosodium salt hydrate was studied.
Other Notes
Degree of hydration varies
Still not finding the right product?
Explore all of our products under Azomethine-H monosodium salt hydrate
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A photochromic thin film based on salicylideneaniline derivatives intercalated layered double hydroxide.
Wang XR, et al.
Chemical Physics Letters, 493(4), 333-339 (2010)