MilliporeSigma
All Photos(1)

286273

Indole-3-acetaldehyde–sodium bisulfite addition compound

Linear Formula:
C10H9NO · NaHSO3
CAS Number:
Molecular Weight:
263.25
MDL number:

SMILES string

[Na].OC(Cc1c[nH]c2ccccc12)S(O)(=O)=O

InChI

1S/C10H11NO4S.Na/c12-10(16(13,14)15)5-7-6-11-9-4-2-1-3-8(7)9;/h1-4,6,10-12H,5H2,(H,13,14,15);/q;+1/p-1

InChI key

WYKPCRCESPDDQX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

may contain excess sodium bisulfite

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service