Documents

288845

Sigma-Aldrich

3-Ethoxybenzamide

98%

Linear Formula:
C2H5OC6H4CONH2
CAS Number:
Molecular Weight:
165.19
MDL number:
PubChem Substance ID:

assay

98%

mp

138-140 °C (lit.)

SMILES string

CCOc1cccc(c1)C(N)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service