289795

Sigma-Aldrich

Potassium tetraborate tetrahydrate

99%

Synonym(s):
Potassium biborate
Linear Formula:
K2B4O7 · 4H2O
CAS Number:
Molecular Weight:
305.50
EC Number:
MDL number:

assay

99%

SMILES string

O.O.O.O.[K+].[K+].[O-]B([O-])OB1OB(OB=O)O1

InChI

1S/B4O7.2K.4H2O/c5-1-8-3-10-4(11-3)9-2(6)7;;;;;;/h;;;4*1H2/q-2;2*+1;;;;

InChI key

FZICZLCGJZRAHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service