290777

Sigma-Aldrich

2-Chloroallyl acetate

97%

Empirical Formula (Hill Notation):
C5H7ClO2
CAS Number:
Molecular Weight:
134.56
MDL number:

assay

97%

refractive index

n20/D 1.437 (lit.)

bp

145 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCC(Cl)=C

Application

Alkylating agent

Signal Word

Danger

Hazard Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2920 3(8) / PGII

WGK Germany

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.