Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

291218

Sigma-Aldrich

Dieldrin

technical grade, ~90%

Synonym(s):

1,2,3,4,10,10-Hexachloro-1,4,4a,5,6,7,8,8a-octahydro-6,7-epoxy-1,4:5,8-dimethanonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6O
CAS Number:
Molecular Weight:
380.91
Beilstein/REAXYS Number:
91396
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

vapor density

13.2 (vs air)

assay

~90%

mp

143-144 °C (lit.)

SMILES string

[H][C@@]12O[C@]1([H])C3CC2[C@]4([H])[C@@]3([H])[C@]5(Cl)C(Cl)=C(Cl)[C@@]4(Cl)C5(Cl)Cl

InChI

1S/C12H8Cl6O/c13-8-9(14)11(16)5-3-1-2(6-7(3)19-6)4(5)10(8,15)12(11,17)18/h2-7H,1H2/t2-,3+,4+,5-,6-,7+,10+,11-

InChI key

DFBKLUNHFCTMDC-PICURKEMSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Snedeker
Environmental health perspectives, 109 Suppl 1, 35-47 (2001-03-17)
Established risk factors for breast cancer explain breast cancer risk only partially. Hence, there has been interest in evaluating what role environmental chemicals, especially those with evidence of being hormonally active agents, play in breast cancer risk. Organochlorine pesticides have
Anumantha G Kanthasamy et al.
Neurotoxicology, 26(4), 701-719 (2005-08-23)
Parkinson's disease (PD) is increasingly recognized as a neurodegenerative disorder strongly associated with environmental chemical exposures. Recent epidemiological data demonstrate that environmental risk factors may play a dominant role as compared to genetic factors in the etiopathogenesis of idiopathic Parkinson's
H J Geyer et al.
Ecotoxicology and environmental safety, 26(1), 45-60 (1993-08-01)
Lipophilic chemicals such as chlorinated hydrocarbon insecticides and other persistent chemicals such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) are fat soluble chemicals and are readily bioconcentrated in animal fat depots. The modifying role of the body fat content in the toxicity of chlorinated
D Damiens et al.
Journal of medical entomology, 50(2), 314-316 (2013-04-02)
The use of the sterile insect technique relies on the release of sterilized mass-reared male insects which, before field releases, endure several unnatural treatments. In the case of Anopheles arabiensis (Patton) sterile insect technique program in Sudan, the genetic background
Harm J Heusinkveld et al.
Environmental science & technology, 46(3), 1842-1848 (2011-12-24)
Current hypotheses link long-term environmental exposure of humans to persistent organochlorine (OC) insecticides lindane (HCH) and dieldrin (HEOD) to the development of neurodegenerative disorders, such as Parkinson's disease. Primary adverse neurological effects of these insecticides are directed at inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service