MilliporeSigma
All Photos(1)

293393

Sigma-Aldrich

Hexanoamide

98%

Synonym(s):
Caproamide
Linear Formula:
CH3(CH2)4CONH2
CAS Number:
Molecular Weight:
115.17
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

98%

mp

100-102 °C (lit.)

SMILES string

CCCCCC(N)=O

InChI

1S/C6H13NO/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H2,7,8)

InChI key

ALBYIUDWACNRRB-UHFFFAOYSA-N

Related Categories

General description

Hexanoamide is the most rapidly hydrolysed substrate for amidase from Aspergillus nidulans. Zirconium mediated conversion of hexanoamide to hexanenitrile has been reported.

Packaging

1 g in glass bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service