Skip to Content
MilliporeSigma

293989

10-DOXYL-nonadecane, free radical

Synonym(s):

4,4-Dimethyl-2,2-dinonyl-3-oxazolidinyloxy, free radical

Sign In to View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C23H47NO2
CAS Number:
Molecular Weight:
369.62
MDL number:
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

refractive index

n20/D 1.459 (lit.)

SMILES string

CCCCCCCCCC1(CCCCCCCCC)OCC(C)(C)N1[O]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service