Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295248

Sigma-Aldrich

1,2-Dichlorotetrafluoroethane

≥99.5%

Synonym(s):

CFC-114

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CF2Cl
CAS Number:
Molecular Weight:
170.92
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

5.93 (vs air)

vapor pressure

1427 mmHg ( 20 °C)

assay

≥99.5%

bp

3.8 °C (lit.)

mp

−94 °C (lit.)

SMILES string

FC(F)(Cl)C(F)(F)Cl

InChI

1S/C2Cl2F4/c3-1(5,6)2(4,7)8

InChI key

DDMOUSALMHHKOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

pictograms

Gas cylinderExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Ozone 1 - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Fang et al.
Anesthesia and analgesia, 84(3), 634-640 (1997-03-01)
Most nonanesthetics (inhaled compounds that neither cause anesthesia when given alone nor decrease the partial pressure of a known inhaled anesthetic required to produce anesthesia) and transitional compounds (inhaled compounds that are less potent than would be predicted by the
L M Dzubow
Journal of the American Academy of Dermatology, 12(5 Pt 1), 796-810 (1985-05-01)
The histologic alterations induced by spray refrigerants independent of and in combination with dermabrasion were studied with the use of the domestic pig as a model. Tissue injury was found to be a function of spray duration and freeze intensity.
R A Strick et al.
The Journal of dermatologic surgery and oncology, 11(12), 1196-1198 (1985-12-01)
Temperatures produced by Cryosthesia -30 degrees C, Cryosthesia -60 degrees C, and Frigiderm were measured in minipigs. Cryosthesia -60 degrees C and Cryosthesia -30 degrees C were both found to rapidly lower skin temperatures to levels that have been shown
Da Yuan et al.
Marine pollution bulletin, 140, 227-240 (2019-02-26)
Spatiotemporal distributions of volatile halogenated organic compounds (VHOCs) were investigated in the marine boundary air and surface seawater of the Changjiang (Yangtze River) estuary and its adjacent East China Sea in two cruises from March 11, 2015 to March 21
V E Maikler
Research in nursing & health, 14(6), 397-403 (1991-12-01)
Although pain management is an important nursing responsibility, few interventions for minimizing distress of infants during painful procedures have been studied. Sixty infants, 2 through 6 months of age, were studied during a routine DPT immunization in order to examine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service