Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

303860

Sigma-Aldrich

cis-Bicyclo[3.3.0]octane-2-carboxylic acid,mixture of endo and exo

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
MDL number:
Pricing and availability is not currently available.

assay

98%

SMILES string

[H][C@@]12[C@@](CC[C@@H]2C(O)=O)([H])CCC1

InChI

1S/C9H14O2/c10-9(11)8-5-4-6-2-1-3-7(6)8/h6-8H,1-5H2,(H,10,11)/t6-,7-,8-/m0/s1

InChI key

AMDLMIANEBVNEY-FXQIFTODSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service