Documents

303941

Sigma-Aldrich

6-Methyl-2-pyridinepropanol

98%

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:

assay

98%

refractive index

n20/D 1.528 (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(CCCO)n1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service