Documents

305162

Sigma-Aldrich

Tris(3-methoxyphenyl)phosphine

97%

Linear Formula:
(CH3OC6H4)3P
CAS Number:
Molecular Weight:
352.36
MDL number:

assay

97%

mp

113-115 °C (lit.)

SMILES string

COc1cccc(c1)P(c2cccc(OC)c2)c3cccc(OC)c3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service