MilliporeSigma
All Photos(1)

305227

Sigma-Aldrich

Phenyl vinylsulfonate

95%

Synonym(s):
Phenyl ethylenesulfonate, Vinylsulfonic acid phenyl ester
Linear Formula:
CH2=CHSO3C6H5
CAS Number:
Molecular Weight:
184.21
Beilstein:
2044718
MDL number:
PubChem Substance ID:
NACRES:
NA.22

assay

95%

bp

44-46 °C/2 mmHg (lit.)

mp

38-42 °C (lit.)

SMILES string

C=CS(=O)(=O)Oc1ccccc1

InChI

1S/C8H8O3S/c1-2-12(9,10)11-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

CILDJVVXNMDAGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phenyl vinylsulfonate is an ester. Palladium complex derived from a tetraphosphine-catalyzed Heck vinylation of phenyl vinylsulfonate was reported. Phenyl vinyl sulfonate reacts with various alkyl furan derivatives to yield the corresponding cycloadducts.

Packaging

1, 5 g in glass bottle

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service