307645

Sigma-Aldrich

Calcium dihydrogenphosphate

technical grade, 85%

Synonym(s):
Calcium bis(dihydrogenphosphate), Calcium phosphate monobasic, prim-Calcium phosphate
Linear Formula:
Ca(H2PO4)2 · H2O
CAS Number:
Molecular Weight:
234.05
EC Number:
MDL number:

grade

technical grade

assay

85%

SMILES string

[Ca++].OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Ca.2H3O4P/c;2*1-5(2,3)4/h;2*(H3,1,2,3,4)/q+2;;/p-2

InChI key

YYRMJZQKEFZXMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

1

Certificate of Analysis
Certificate of Origin
Caroline Öhman et al.
Journal of materials science. Materials in medicine, 26(7), 210-210 (2015-07-15)
The porosity of calcium phosphate cements has an impact on several important parameters, such as strength, resorbability and bioactivity. A model to predict the porosity for biomedical cements would hence be a useful tool. At the moment such a model...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.