307971

Sigma-Aldrich

2-Ethylbutyl acetate

98%

Linear Formula:
CH3CO2CH2CH(C2H5)2
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

refractive index

n20/D 1.41 (lit.)

bp

160 °C/740 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CCC(CC)COC(C)=O

InChI

1S/C8H16O2/c1-4-8(5-2)6-10-7(3)9/h8H,4-6H2,1-3H3

InChI key

HQLKZWRSOHTERR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

RIDADR

UN1177 - class 3 - PG 3 - 2-Ethylbutyl acetate

WGK Germany

3

Flash Point(F)

127.4 °F

Flash Point(C)

53 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.