Skip to Content
MilliporeSigma

311065

Hexanophenone

≥99%

Synonym(s):

Amyl phenyl ketone

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C6H5CO(CH2)4CH3
CAS Number:
Molecular Weight:
176.25
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
213-394-6
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Hexanophenone, ≥99%

InChI key

MAHPVQDVMLWUAG-UHFFFAOYSA-N

InChI

1S/C12H16O/c1-2-3-5-10-12(13)11-8-6-4-7-9-11/h4,6-9H,2-3,5,10H2,1H3

SMILES string

CCCCCC(=O)c1ccccc1

assay

≥99%

refractive index

n20/D 1.5105 (lit.)

bp

265 °C (lit.)

mp

25-26 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

HPLC standard

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak
Martin Gilar et al.
Journal of chromatography. A, 1381, 110-117 (2015-01-22)
Sample introduction in microfluidic liquid chromatography often generates wide zones rather than peaks, especially when a large sample volume (relative to column volume) is injected. Formation of wide injection zones can be further amplified when the sample is dissolved in
Martin Gilar et al.
Journal of chromatography. A, 1390, 86-94 (2015-03-10)
Chromatographic zone broadening is a common issue in microfluidic chromatography, where the sample volume introduced on column often exceeds the column void volume. To better understand the propagation of wide chromatographic zones on a separation device, a series of MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service