316741

Sigma-Aldrich

(±)-Epinephrine

95%

Synonym(s):
(±)-Adrenalin, 4-[1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol, (±)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, 3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol
Linear Formula:
(HO)2C6H3CH(CH2NHCH3)OH
CAS Number:
Molecular Weight:
183.20
EC Number:
MDL number:

assay

95%

mp

197 °C (dec.) (lit.)

SMILES string

CNCC(O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3

InChI key

UCTWMZQNUQWSLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Adrenoceptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2811 6.1 / PGII

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.