Documents

323292

Sigma-Aldrich

(S)-(−)-Propranolol hydrochloride

98%

Synonym(s):
(S)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride
Linear Formula:
C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl
CAS Number:
Molecular Weight:
295.80
Beilstein/REAXYS Number:
3574966
EC Number:
MDL number:

assay

98%

optical activity

[α]20/D −26°, c = 1 in ethanol

mp

193-195 °C (lit.)

SMILES string

Cl[H].CC(C)NC[C@H](O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H/t14-;/m0./s1

InChI key

ZMRUPTIKESYGQW-UQKRIMTDSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service