326658

Sigma-Aldrich

Palladium

powder, 1.0-1.5 μm, ≥99.9% trace metals basis

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99.9% trace metals basis

form

powder

resistivity

9.96 μΩ-cm, 20°C

particle size

1.0-1.5 μm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Bin Kim et al.
Journal of nanoscience and nanotechnology, 13(1), 517-522 (2013-05-08)
In order to form flexible printed circuits through inkjet printing technique, the Pd(ll) catalyst ink was printed on the surface of polyimide film modified with KOH solution and then reduced with NaBH4 solution to extract the Pd(O) catalyst nuclei. The...
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.
Hyunsu Kim et al.
Journal of nanoscience and nanotechnology, 13(1), 533-536 (2013-05-08)
ZnSnO3 one-dimensional (1D) strutures were synthesized by using an evaporation technique. The morphology, crystal structure, and enhanced sensing properties of the ZnSnO3 structures functionalized with Pd to CO gas at 300 degrees C were investigated. The diameters of the 1D...
Z Y Bian et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(8), 1873-1879 (2013-04-13)
A Pd/C gas-diffusion cathode which generated H2O2 through a two-electron reduction process of fed oxygen molecule was used to degrade 4-chlorophenol in an undivided electrolysis device. The kinetics of 4-chlorophenol degradation has been investigated by the electrochemical oxidation processes. By...
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service