Documents

330701

Sigma-Aldrich

2,5,5-Tris(trimethylsilyl)-1,3-cyclopentadiene

99%

Empirical Formula (Hill Notation):
C14H30Si3
CAS Number:
Molecular Weight:
282.64
MDL number:

assay

99%

refractive index

n20/D 1.473 (lit.)

density

0.841 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C1=CC(C=C1)([Si](C)(C)C)[Si](C)(C)C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service