332941

Sigma-Aldrich

1-Bromodocosane

96%

Synonym(s):
Docosyl bromide
Linear Formula:
CH3(CH2)21Br
CAS Number:
Molecular Weight:
389.50
Beilstein/REAXYS Number:
1770458
EC Number:
MDL number:
PubChem Substance ID:

assay

96%

bp

225 °C/0.6 mmHg (lit.)

mp

44-46 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCBr

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service