Documents

333077

Sigma-Aldrich

Dimethyl (4-nitrobenzylidene)malonate

99%

Linear Formula:
O2NC6H4CH=C(CO2CH3)2
CAS Number:
Molecular Weight:
265.22
MDL number:

assay

99%

mp

137-139 °C (lit.)

SMILES string

COC(=O)\C(=C\c1ccc(cc1)[N+]([O-])=O)C(=O)OC

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service