335185

Sigma-Aldrich

Methyl stearate

99%

Synonym(s):
Methyl octadecanoate, Stearic acid methyl ester
Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein/REAXYS Number:
1786213
EC Number:
MDL number:

assay

99%

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service