336300

Sigma-Aldrich

Tungsten boride

99%, −325 mesh

Linear Formula:
WB
CAS Number:
Molecular Weight:
194.65
EC Number:
MDL number:

assay

99%

particle size

−325 mesh

SMILES string

B#[W]

InChI

1S/B.W

InChI key

JEEHQNXCPARQJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service