Skip to Content
MilliporeSigma

337625

Bromo-tert-butyl(methoxy)phenylsilane

95%

Synonym(s):

tert-Butyl(methoxy)phenylbromosilane, TBMPSiBr

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C11H17BrOSi
CAS Number:
Molecular Weight:
273.24
UNSPSC Code:
12352000
MDL number:
Beilstein/REAXYS Number:
4983916

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

CO[Si](Br)(c1ccccc1)C(C)(C)C

assay

95%

density

1.218 g/mL at 20 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service